1,3-Bis(4-chlorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4529-1G
B4529-5G
TCB4529-1GEA
153.26
CAD
TCB4529-1G
TCB4529-5G
1,3-Bis(4-chlorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis(4-chlorophenyl)urea
CAS Number: 1219-99-4
MDL Number: MFCD00018541
Molecular Formula: C13H10Cl2N2O
Molecular Weight: 281.14
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 306
Formula:
C₁₃H₁₀Cl₂N₂O MW: 281.14 g/mol Melting Pt: 306 °C |
MDL Number:
MFCD00018541 CAS Number: 1219-99-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...