Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate ≥98.0% (by HPLC)

Supplier: TCI America

B4524-1G B4524-200MG
TCB4524-1GEA 276.03 CAD
TCB4524-1G TCB4524-200MG
Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate ≥98.0% (by HPLC)
Bis(4-bromophenylboronic acid)scylloinositol complex dipotassium tetrahydrate

CAS Number: 1537876-29-1
Molecular Formula: C18H14B2Br2K2O6
Molecular Weight: 585.93
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₈H₁₄B₂Br₂K₂O₆
CAS Number: 1537876-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=457f614dacc0c3b169a9a2ac23404bf45bc480cc&vid=457f614dacc0c3b169a9a2ac23404bf45bc480cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR