Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
Supplier: TCI America
B4523-1G
B4523-200MG
TCB4523-1GEA
243
CAD
TCB4523-1G
TCB4523-200MG
Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
Bis(3-bromophenylboronic acid)scyllo inositol complex dipotassium pentahydrate
CAS Number: 1537876-31-5
Molecular Formula: C18H14B2Br2K2O6
Molecular Weight: 585.93
Form: Crystal
Color: White
Formula:
C₁₈H₁₄B₂Br₂K₂O₆ |
CAS Number:
1537876-31-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...