Benzo[b]furan-3-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Benzofuran-3-ylboronic acid

B4522-1G B4522-5G
TCB4522-1GEA 201.61 CAD
TCB4522-1G TCB4522-5G
Benzo[b]furan-3-boronic acid (contains varying amounts of Anhydride)
Benzo[b]furan-3-boronic acid

CAS Number: 317830-83-4
MDL Number: MFCD06801687
Molecular Formula: C8H7BO3
Molecular Weight: 161.95
Form: Crystal
Color: Very Pale Yellow

Formula: C₈H₇BO₃
MW: 161.95 g/mol
MDL Number: MFCD06801687
CAS Number: 317830-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105109eab619e08169dfecd65ce908d145adcf40&vid=105109eab619e08169dfecd65ce908d145adcf40&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR