1,3-Bis[4-(trifluoromethyl)phenyl]urea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1,3-Bis(4-(trifluoromethyl)phenyl)urea

B4518-1G B4518-200MG
TCB4518-1GEA 730.59 CAD
TCB4518-1G TCB4518-200MG
1,3-Bis[4-(trifluoromethyl)phenyl]urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis[4-(trifluoromethyl)phenyl]urea

CAS Number: 1960-88-9
MDL Number: MFCD01908917
Molecular Formula: C15H10F6N2O
Molecular Weight: 348.25
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 227
Lambda max.: 264 nm (EtOH)

Formula: C₁₅H₁₀F₆N₂O
MW: 348.25 g/mol
Melting Pt: 227 °C
MDL Number: MFCD01908917
CAS Number: 1960-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00a95bf52a23a14e400e2bc7717c7f489c897b71&vid=00a95bf52a23a14e400e2bc7717c7f489c897b71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR