1,3-Bis(4-fluorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4517-1G
B4517-5G
TCB4517-1GEA
145.36
CAD
TCB4517-1G
TCB4517-5G
1,3-Bis(4-fluorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis(4-fluorophenyl)urea
CAS Number: 370-22-9
MDL Number: MFCD00017925
Molecular Formula: C13H10F2N2O
Molecular Weight: 248.23
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 267
Formula:
C₁₃H₁₀F₂N₂O Melting Pt: 267 °C |
MDL Number:
MFCD00017925 CAS Number: 370-22-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...