2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥98.0%

Supplier: TCI America

B4512-1G B4512-5G
TCB4512-1GEA 58.46 CAD
TCB4512-1G TCB4512-5G
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥98.0%
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine

CAS Number: 49669-14-9
MDL Number: MFCD12406888
Molecular Formula: C9H10BrNO2
Molecular Weight: 244.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45

Formula: C₉H₁₀BrNO₂
Melting Pt: 45 °C
MDL Number: MFCD12406888
CAS Number: 49669-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=829b67da25eac21e0b63f9949c1b27f616d92b4a&vid=829b67da25eac21e0b63f9949c1b27f616d92b4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR