4-Bromo-3-(trifluoromethyl)phenol ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-5-hydroxybenzotrifluoride

B4494-1G B4494-5G
TCB4494-1GEA 37.92 CAD
TCB4494-1G TCB4494-5G
4-Bromo-3-(trifluoromethyl)phenol ≥98.0%
4-Bromo-3-(trifluoromethyl)phenol

CAS Number: 320-49-0
MDL Number: MFCD03095342
Molecular Formula: C7H4BrF3O
Molecular Weight: 241.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 48
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₇H₄BrF₃O
Melting Pt: 48 °C
Flash Pt: 110 °C
MDL Number: MFCD03095342
CAS Number: 320-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24571315e9a016695ad35033b7374b7f75c7b8bb&vid=24571315e9a016695ad35033b7374b7f75c7b8bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR