4-Bromo-2-(trifluoromethyl)phenol ≥97.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-hydroxybenzotrifluoride

B4493-25G B4493-5G
TCB4493-25GEA 374.46 CAD
TCB4493-25G TCB4493-5G
4-Bromo-2-(trifluoromethyl)phenol ≥97.0%
4-Bromo-2-(trifluoromethyl)phenol

CAS Number: 50824-04-9
MDL Number: MFCD05668981
Molecular Formula: C7H4BrF3O
Molecular Weight: 241.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85

Formula: C₇H₄BrF₃O
Melting Pt: 85 °C
MDL Number: MFCD05668981
CAS Number: 50824-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=782ca54e88a1f8e37ea59e8c19492f6b2af849f0&vid=782ca54e88a1f8e37ea59e8c19492f6b2af849f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR