1,3-Bis(4-methoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4483-1G B4483-5G
TCB4483-1GEA 248.06 CAD
TCB4483-1G TCB4483-5G
1,3-Bis(4-methoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis(4-methoxyphenyl)urea

CAS Number: 1227-44-7
MDL Number: MFCD00025783
Molecular Formula: C15H16N2O3
Molecular Weight: 272.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 241

Formula: C₁₅H₁₆N₂O₃
Melting Pt: 241 °C
MDL Number: MFCD00025783
CAS Number: 1227-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81ba8df00eac85664cf8e87327ecaf1fabae5d09&vid=81ba8df00eac85664cf8e87327ecaf1fabae5d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR