6-Bromoquinoxaline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4476-1G B4476-5G
TCB4476-1GEA 94.8 CAD
TCB4476-1G TCB4476-5G
6-Bromoquinoxaline ≥98.0% (by GC, titration analysis)
6-Bromoquinoxaline

CAS Number: 50998-17-9
MDL Number: MFCD00837757
Molecular Formula: C8H5BrN2
Molecular Weight: 209.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 149
Melting point (°C): 56
Flash Point (°C): 110
Lambda max.: 322 nm (H2O)

Formula: C₈H₅BrN₂
Boiling Pt: 149 °C
Melting Pt: 56 °C
Flash Pt: 110 °C
MDL Number: MFCD00837757
CAS Number: 50998-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64f59095a9e89189895c49463bd1b56e0a3f32ac&vid=64f59095a9e89189895c49463bd1b56e0a3f32ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR