9-(4-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)

Supplier: TCI America

B4475-1G B4475-200MG
TCB4475-1GEA 314.1 CAD
TCB4475-1G TCB4475-200MG
9-(4-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
9-(4-Bromophenyl)-10-phenylanthracene

CAS Number: 625854-02-6
MDL Number: MFCD24842536
Molecular Formula: C26H17Br
Molecular Weight: 409.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 227

Formula: C₂₆H₁₇Br
MW: 409.33 g/mol
Melting Pt: 227 °C
MDL Number: MFCD24842536
CAS Number: 625854-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bbd4782c7307987d109fc883ad8c48dbec0c93a&vid=5bbd4782c7307987d109fc883ad8c48dbec0c93a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR