trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: trans,trans-Bis[4-(trifluoromethyl)benzal]acetone, trans,trans-Bis[4-(trifluoromethyl)benzylidene]acetone, (1E,4E)-1,5-Bis(4-(trifluoromethyl)phenyl)penta-1,4-dien-3-one, trans,trans-Bis[4-(trifluoromethyl)styryl] Ketone

B4468-1G B4468-200MG
TCB4468-1GEA 431.34 CAD
TCB4468-1G TCB4468-200MG
trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one ≥98.0% (by HPLC)
trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one

CAS Number: 103836-71-1
Molecular Formula: C19H12F6O
Molecular Weight: 370.29
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Lambda max.: 388 nm (THF)


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Ligands, Fluorinated Building Blocks
Other Product Names / Synonyms: trans,trans-Bis[4-(trifluoromethyl)benzal]acetone, trans,trans-Bis[4-(trifluoromethyl)benzylidene]acetone, trans,trans-Bis[4-(trifluoromethyl)styryl] Ketone

Formula: C₁₉H₁₂F₆O
MW: 370.29 g/mol
MDL Number: MFCD05662426
CAS Number: 103836-71-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR