trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0%

Supplier: TCI America

Synonyms: trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxystyryl) Ketone

B4467-1G B4467-200MG
TCB4467-1GEA 294.2 CAD
TCB4467-1G TCB4467-200MG
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0%
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one

CAS Number: 37951-12-5
MDL Number: MFCD00014901
Molecular Formula: C19H18O3
Molecular Weight: 294.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129
Lambda max.: 357 nm (THF)


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Ligands
Other Product Names / Synonyms: trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxystyryl) Ketone

Formula: C₁₉H₁₈O₃
MW: 294.35 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00014901
CAS Number: 37951-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f79ce526a88a4b3e75990c21ae7024b3a3d2a3d4&vid=f79ce526a88a4b3e75990c21ae7024b3a3d2a3d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR