5-Bromo-2-methoxy-3-nitropyridine ≥96.0% (by GC)

Supplier: TCI America

B4461-1G B4461-5G
TCB4461-1GEA 56.88 CAD
TCB4461-1G TCB4461-5G
5-Bromo-2-methoxy-3-nitropyridine ≥96.0% (by GC)
5-Bromo-2-methoxy-3-nitropyridine

CAS Number: 152684-30-5
MDL Number: MFCD07374968
Molecular Formula: C6H5BrN2O3
Molecular Weight: 233.02
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 90

Formula: C₆H₅BrN₂O₃
Melting Pt: 90 °C
MDL Number: MFCD07374968
CAS Number: 152684-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef6bba662797fd28e02d2410a8f1cb52e27ca8f8&vid=ef6bba662797fd28e02d2410a8f1cb52e27ca8f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR