5-Bromo-2-methox-6-methylypyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine

B4460-1G B4460-5G
TCB4460-1GEA 34.76 CAD
TCB4460-1G TCB4460-5G
5-Bromo-2-methox-6-methylypyridine ≥98.0%
5-Bromo-2-methox-6-methylypyridine

CAS Number: 126717-59-7
MDL Number: MFCD03094943
Molecular Formula: C7H8BrNO
Molecular Weight: 202.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 106
Specific Gravity (20/20): 1.48

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 86 °C
Density: 1.48 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD03094943
CAS Number: 126717-59-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=846e9e618cd595c8c2eb920068198067b2038350&vid=846e9e618cd595c8c2eb920068198067b2038350&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR