2-Bromodibenzofuran ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromodibenzo[b,d]furan

B4459-1G B4459-5G
TCB4459-1GEA 87.53 CAD
TCB4459-1G TCB4459-5G
2-Bromodibenzofuran ≥98.0% (by GC)
2-Bromodibenzofuran

CAS Number: 86-76-0
MDL Number: MFCD00092338
Molecular Formula: C12H7BrO
Molecular Weight: 247.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 112

Formula: C₁₂H₇BrO
MW: 247.09 g/mol
Boiling Pt: 220 °C
Melting Pt: 112 °C
MDL Number: MFCD00092338
CAS Number: 86-76-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR