2-Bromo-3-decylthiophene ≥97.0%
Supplier: TCI America
B4455-25G
B4455-5G
TCB4455-25GEA
543.84
CAD
TCB4455-25G
TCB4455-5G
2-Bromo-3-decylthiophene ≥97.0%
2-Bromo-3-decylthiophene
CAS Number: 144012-09-9
MDL Number: MFCD16619251
Molecular Formula: C14H23BrS
Molecular Weight: 303.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.16
Formula:
C₁₄H₂₃BrS Density: 1.16 (20 °C) |
MDL Number:
MFCD16619251 CAS Number: 144012-09-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...