3-(4-Bromophenyl)-9-phenylcarbazole ≥98.0% (by GC)

Supplier: TCI America

B4452-1G B4452-5G
TCB4452-1GEA 47.4 CAD
TCB4452-1G TCB4452-5G
3-(4-Bromophenyl)-9-phenylcarbazole ≥98.0% (by GC)
3-(4-Bromophenyl)-9-phenylcarbazole

CAS Number: 1028647-93-9
MDL Number: MFCD14582939
Molecular Formula: C24H16BrN
Molecular Weight: 398.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138

Formula: C₂₄H₁₆BrN
Melting Pt: 138 °C
MDL Number: MFCD14582939
CAS Number: 1028647-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89b7b067631a3a617ca52c991caee6aacdea2c64&vid=89b7b067631a3a617ca52c991caee6aacdea2c64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR