2-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-9,9-diphenyl-9H-fluorene

B4450-1G B4450-5G
TCB4450-1GEA 52.93 CAD
TCB4450-1G TCB4450-5G
2-Bromo-9,9-diphenylfluorene ≥98.0% (by GC)
2-Bromo-9,9-diphenylfluorene

CAS Number: 474918-32-6
MDL Number: MFCD09907957
Molecular Formula: C25H17Br
Molecular Weight: 397.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 219

Formula: C₂₅H₁₇Br
MW: 397.32 g/mol
Melting Pt: 219 °C
MDL Number: MFCD09907957
CAS Number: 474918-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=528aa1ef6dfe0991b6b298e669ec6639958f5bad&vid=528aa1ef6dfe0991b6b298e669ec6639958f5bad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR