4-Bromodibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromodibenzo[b,d]thiophene

B4449-1G B4449-5G
TCB4449-1GEA 48.98 CAD
TCB4449-1G TCB4449-5G
4-Bromodibenzothiophene ≥98.0% (by GC)
4-Bromodibenzothiophene

CAS Number: 97511-05-2
MDL Number: MFCD02683746
Molecular Formula: C12H7BrS
Molecular Weight: 263.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87

Formula: C₁₂H₇BrS
MW: 263.15 g/mol
Melting Pt: 87 °C
MDL Number: MFCD02683746
CAS Number: 97511-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a730fa2928f996e328192709b18772b5a896021&vid=0a730fa2928f996e328192709b18772b5a896021&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR