6-Bromo-2-chlorobenzothiazole ≥96.0%
Supplier: TCI America
B4448-1G
B4448-5G
TCB4448-1GEA
79
CAD
TCB4448-1G
TCB4448-5G
6-Bromo-2-chlorobenzothiazole ≥96.0%
6-Bromo-2-chlorobenzothiazole
CAS Number: 80945-86-4
MDL Number: MFCD04971822
Molecular Formula: C7H3BrClNS
Molecular Weight: 248.52
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 100
Formula:
C₇H₃BrClNS Boiling Pt: 157 °C Melting Pt: 100 °C |
MDL Number:
MFCD04971822 CAS Number: 80945-86-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...