(S)-1-Boc-3-hydroxypiperidine ≥98.0%

Supplier: TCI America

Synonyms: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

B4446-1G B4446-5G
TCB4446-1GEA 34.76 CAD
TCB4446-1G TCB4446-5G
(S)-1-Boc-3-hydroxypiperidine ≥98.0%
(S)-1-Boc-3-hydroxypiperidine

CAS Number: 143900-44-1
MDL Number: MFCD04115306
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 110
Specific rotation [a]20/D: 22 deg (C=5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1-Boc-3-hydroxypiperidine, (S)-1-Boc-3-piperidinol

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD04115307
CAS Number: 143900-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6027812b8b073ec92b52d1817cfe03c1de06233&vid=c6027812b8b073ec92b52d1817cfe03c1de06233&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR