2-Bromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

B4439-1G B4439-5G
TCB4439-1GEA 61.15 CAD
TCB4439-1G TCB4439-5G
2-Bromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
2-Bromo-9-phenylcarbazole

CAS Number: 94994-62-4
MDL Number: MFCD24387061
Molecular Formula: C18H12BrN
Molecular Weight: 322.21
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 90

Formula: C₁₈H₁₂BrN
Melting Pt: 90 °C
MDL Number: MFCD24387061
CAS Number: 94994-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=318b6c150009585411b133913eb999fad9844966&vid=318b6c150009585411b133913eb999fad9844966&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR