5-Bromo-2-benzoxazolinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromo-2-hydroxybenzoxazole, 5-Bromo-2-benzoxazolone, 5-Bromobenzo[d]oxazol-2(3H)-one

B4438-1G B4438-5G
TCB4438-1GEA 86.9 CAD
TCB4438-1G TCB4438-5G
5-Bromo-2-benzoxazolinone ≥98.0% (by GC, titration analysis)
5-Bromo-2-benzoxazolinone

CAS Number: 14733-73-4
MDL Number: MFCD01664246
Molecular Formula: C7H4BrNO2
Molecular Weight: 214.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 217


Other Product Names / Synonyms: 5-Bromo-2-benzoxazolone, 5-Bromo-2-hydroxybenzoxazole

Formula: C₇H₄BrNO₂
MW: 214.02 g/mol
Melting Pt: 217 °C
MDL Number: MFCD01664246
CAS Number: 14733-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fbd08c8c5506d0c26ba80fb68378143cd028429&vid=8fbd08c8c5506d0c26ba80fb68378143cd028429&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR