tert-Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B4434-1G B4434-5G
TCB4434-1GEA 156.42 CAD
TCB4434-1G TCB4434-5G
tert-Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
tert-Butylamine hydroiodide

CAS Number: 39557-45-4
Molecular Formula: C4H11N
Molecular Weight: 201.05
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Melting point (°C): 221

Formula: C₄H₁₁N
Melting Pt: 221 °C
CAS Number: 39557-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0716936925f4d4c42ccc3b0dc374f8657469d3b&vid=b0716936925f4d4c42ccc3b0dc374f8657469d3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR