Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B4433-1G B4433-5G
TCB4433-1GEA 145.36 CAD
TCB4433-1G TCB4433-5G
Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Butylamine hydroiodide

CAS Number: 36945-08-1
Molecular Formula: C4H11N
Molecular Weight: 201.05
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Melting point (°C): 171

Formula: C₄H₁₁N
Melting Pt: 171 °C
CAS Number: 36945-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e156000b068b841a518b910d16b98422293540fe&vid=e156000b068b841a518b910d16b98422293540fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR