2-Bromo-7-iodofluorene ≥98.0% (by GC)

Supplier: TCI America

B4431-25G B4431-5G
TCB4431-25GEA 277.76 CAD
TCB4431-25G TCB4431-5G
2-Bromo-7-iodofluorene ≥98.0% (by GC)
2-Bromo-7-iodofluorene

CAS Number: 123348-27-6
MDL Number: MFCD12024276
Molecular Formula: C13H8BrI
Molecular Weight: 371.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 184

Formula: C₁₃H₈BrI
Melting Pt: 184 °C
MDL Number: MFCD12024276
CAS Number: 123348-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d55c3a50131fe52b886bd00a6eeafd3b8aaad157&vid=d55c3a50131fe52b886bd00a6eeafd3b8aaad157&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR