2,2-Bis(3-amino-4-hydroxylphenyl)propane ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4,4'-Isopropylidenebis(2-aminophenol), 4,4'-(Propane-2,2-diyl)bis(2-aminophenol)
B4429-25G
B4429-5G
TCB4429-25GEA
388.68
CAD
TCB4429-25G
TCB4429-5G
2,2-Bis(3-amino-4-hydroxylphenyl)propane ≥97.0% (by HPLC, titration analysis)
2,2-Bis(3-amino-4-hydroxylphenyl)propane
CAS Number: 1220-78-6
MDL Number: MFCD00437413
Molecular Formula: C15H18N2O2
Molecular Weight: 258.32
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 254
Product Categories: Materials Chemistry, Polyimide Raw Materials
Other Product Names / Synonyms: 4,4'-Isopropylidenebis(2-aminophenol)
Formula:
C₁₅H₁₈N₂O₂ MW: 258.32 g/mol Melting Pt: 254 °C |
MDL Number:
MFCD00437413 CAS Number: 1220-78-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...