1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4428-25G B4428-5G
TCB4428-25GEA 328.96 CAD
TCB4428-25G TCB4428-5G
1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene ≥98.0% (by HPLC, titration analysis)
1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene

CAS Number: 94525-05-0
MDL Number: MFCD07368071
Molecular Formula: C20H14F6N2O2
Molecular Weight: 428.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 134

Formula: C₂₀H₁₄F₆N₂O₂
Melting Pt: 134 °C
MDL Number: MFCD07368071
CAS Number: 94525-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39231c320b63fe1fe140e87f7f755aee4c6419ac&vid=39231c320b63fe1fe140e87f7f755aee4c6419ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR