9-(3-Bromophenyl)carbazole ≥98.0% (by GC)

Supplier: TCI America

B4427-1G B4427-5G
TCB4427-1GEA 52.93 CAD
TCB4427-1G TCB4427-5G
9-(3-Bromophenyl)carbazole ≥98.0% (by GC)
9-(3-Bromophenyl)carbazole

CAS Number: 185112-61-2
MDL Number: MFCD20486475
Molecular Formula: C18H12BrN
Molecular Weight: 322.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69

Formula: C₁₈H₁₂BrN
Melting Pt: 69 °C
MDL Number: MFCD20486475
CAS Number: 185112-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=119f014fbeb3a5988557061eda5619a0d7c64e59&vid=119f014fbeb3a5988557061eda5619a0d7c64e59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR