9-Bromo-10-(2-naphthyl)anthracene ≥98.0% (by GC)

Supplier: TCI America

B4426-1G B4426-5G
TCB4426-1GEA 92.59 CAD
TCB4426-1G TCB4426-5G
9-Bromo-10-(2-naphthyl)anthracene ≥98.0% (by GC)
9-Bromo-10-(2-naphthyl)anthracene

CAS Number: 474688-73-8
MDL Number: MFCD09832882
Molecular Formula: C24H15Br
Molecular Weight: 383.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 171

Formula: C₂₄H₁₅Br
MW: 383.29 g/mol
Melting Pt: 171 °C
MDL Number: MFCD09832882
CAS Number: 474688-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fb18dad93742aeb58d83e32feece7a2b1f59e1a&vid=6fb18dad93742aeb58d83e32feece7a2b1f59e1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR