2-(4-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0%
Supplier: TCI America
Synonyms:
N-([1,1'-Biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine
B4424-1G
B4424-5G
TCB4424-1GEA
77.74
CAD
TCB4424-1G
TCB4424-5G
2-(4-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0%
2-(4-Biphenylyl)amino-9,9-dimethylfluorene
CAS Number: 897671-69-1
MDL Number: MFCD16036286
Molecular Formula: C27H23N
Molecular Weight: 361.49
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 147
Formula:
C₂₇H₂₃N MW: 361.49 g/mol Melting Pt: 147 °C |
MDL Number:
MFCD16036286 CAS Number: 897671-69-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...