2-Bromo-5-nitrothiazole ≥97.0% (by GC)

Supplier: TCI America

B4413-1G B4413-5G
TCB4413-1GEA 75.84 CAD
TCB4413-1G TCB4413-5G
2-Bromo-5-nitrothiazole ≥97.0% (by GC)
2-Bromo-5-nitrothiazole

CAS Number: 3034-48-8
MDL Number: MFCD00005317
Molecular Formula: C3HBrN2O2S
Molecular Weight: 209.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85

Formula: C₃HBrN₂O₂S
Melting Pt: 85 °C
MDL Number: MFCD00005317
CAS Number: 3034-48-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59a21d80b149d33ffcaec4c6eaed87ed1e8dbee0&vid=59a21d80b149d33ffcaec4c6eaed87ed1e8dbee0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR