6-Bromo-2-hydroxypyridine ≥98.0%
Supplier: TCI America
Synonyms:
6-Bromo-2-pyridone, 2-Bromo-6-hydroxypyridine, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridinol
B4408-1G
B4408-5G
TCB4408-1GEA
63.2
CAD
TCB4408-1G
TCB4408-5G
6-Bromo-2-hydroxypyridine ≥98.0%
6-Bromo-2-hydroxypyridine
CAS Number: 27992-32-1
MDL Number: MFCD00234043
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123
Other Product Names / Synonyms: 6-Bromo-2-pyridinol, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridone
Formula:
C₅H₄BrNO MW: 174 Melting Pt: 123 °C |
MDL Number:
MFCD00234043 CAS Number: 27992-32-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...