2-Bromo-4-chloropyridine ≥98.0% (by GC)
Supplier: TCI America
B4407-1G
B4407-5G
TCB4407-1GEA
47.87
CAD
TCB4407-1G
TCB4407-5G
2-Bromo-4-chloropyridine ≥98.0% (by GC)
2-Bromo-4-chloropyridine
CAS Number: 22918-01-0
MDL Number: MFCD03840757
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 27
Flash Point (°C): 110
Formula:
C₅H₃BrClN Boiling Pt: 85 °C Melting Pt: 27 °C Flash Pt: 110 °C |
MDL Number:
MFCD03840757 CAS Number: 22918-01-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...