1-Bromo-3,7-dimethyloctane ≥93.0% (by GC)

Supplier: TCI America

B4401-25G B4401-5G
TCB4401-25GEA 1008.36 CAD
TCB4401-25G TCB4401-5G
1-Bromo-3,7-dimethyloctane ≥93.0% (by GC)
1-Bromo-3,7-dimethyloctane

CAS Number: 3383-83-3
MDL Number: MFCD00039201
Molecular Formula: C10H21Br
Molecular Weight: 221.18
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 99
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 3,7-Dimethyloctyl Bromide

Formula: C₁₀H₂₁Br
Boiling Pt: 217 °C
Density: 1.06 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00039201
CAS Number: 3383-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f8a5f7ae74784770d64c8453cdb76e8c26f305e&vid=9f8a5f7ae74784770d64c8453cdb76e8c26f305e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR