1-(1-Boc-piperidino)-4-bromo-1H-pyrazole ≥98.0%
Supplier: TCI America
Synonyms:
tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole
CAS Number: 877399-50-3
MDL Number: MFCD09831980
Molecular Formula: C13H20BrN3O2
Molecular Weight: 330.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 79
Other Product Names / Synonyms: 4-(4-Bromopyrazol-1-yl)piperidine-1-carboxylic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl-4-(4-bromopyrazol-1-yl)piperidine, 1-Boc-4-(4-bromopyrazol-1-yl)piperidine
Formula:
C₁₃H₂₀BrN₃O₂ MW: 330.23 g/mol Melting Pt: 79 °C |
MDL Number:
MFCD09831980 CAS Number: 877399-50-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...