(4-Bromophenyl)acetone ≥98.0% (by GC)
Supplier: TCI America
B4387-25G
B4387-5G
TCB4387-25GEA
348.71
CAD
TCB4387-25G
TCB4387-5G
(4-Bromophenyl)acetone ≥98.0% (by GC)
(4-Bromophenyl)acetone
CAS Number: 6186-22-7
MDL Number: MFCD00210401
Molecular Formula: C9H9BrO
Molecular Weight: 213.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Specific Gravity (20/20): 1.42
Other Product Names / Synonyms: 1-(4-Bromophenyl)-2-propanone
Formula:
C₉H₉BrO Boiling Pt: 139 °C Density: 1.42 (20 °C) |
MDL Number:
MFCD00210401 CAS Number: 6186-22-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...