Bis(2-hydroxyethyl)dimethylammonium chloride ≥98.0%
Supplier: TCI America
B4384-25G
B4384-5G
TCB4384-25GEA
290.72
CAD
TCB4384-25G
TCB4384-5G
Bis(2-hydroxyethyl)dimethylammonium chloride ≥98.0%
Bis(2-hydroxyethyl)dimethylammonium chloride
CAS Number: 38402-02-7
MDL Number: MFCD00041987
Molecular Formula: C6H16ClNO2
Molecular Weight: 169.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₆H₁₆ClNO₂ |
MDL Number:
MFCD00041987 CAS Number: 38402-02-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...