2-Bromocarbazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-9H-carbazole

B4381-5G B4381-25G
TCB4381-5GEA 77.42 CAD
TCB4381-5G TCB4381-25G
2-Bromocarbazole ≥98.0% (by GC)
2-Bromocarbazole

CAS Number: 3652-90-2
MDL Number: MFCD09750430
Molecular Formula: C12H8BrN
Molecular Weight: 246.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 255

Formula: C₁₂H₈BrN
MW: 246.11 g/mol
Melting Pt: 255 °C
MDL Number: MFCD09750430
CAS Number: 3652-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39f3c7168f858c2f35f137cac4404f5389eb860d&vid=39f3c7168f858c2f35f137cac4404f5389eb860d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR