4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene ≥95.0% (by GC)
Supplier: TCI America
B4380-1G
B4380-5G
TCB4380-1GEA
196.71
CAD
TCB4380-1G
TCB4380-5G
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene ≥95.0% (by GC)
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-S-indacene
CAS Number: 960079-28-1
Molecular Formula: C28H45Br
Molecular Weight: 461.57
Purity/Analysis Method: >95.0% (GC)
Form: Crystalline Powder
Melting point (°C): 155
Other Product Names / Synonyms: Eind-Br
Formula:
C₂₈H₄₅Br Melting Pt: 155 °C |
CAS Number:
960079-28-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...