4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene ≥95.0% (by GC)

Supplier: TCI America

B4379-1G B4379-5G
TCB4379-1GEA 137.46 CAD
TCB4379-1G TCB4379-5G
4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene ≥95.0% (by GC)
4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene

CAS Number: 1142818-90-3
Molecular Formula: C24H37Br
Molecular Weight: 405.46
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: EMind-Br

Formula: C₂₄H₃₇Br
Melting Pt: 110 °C
CAS Number: 1142818-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=794352a56e414584b7cf8b9961b28c90a2248f3d&vid=794352a56e414584b7cf8b9961b28c90a2248f3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR