1-(2-Bromoethoxy)-4-fluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Fluorophenoxy-ethylbromide

B4376-25G B4376-5G
TCB4376-25GEA 692.04 CAD
TCB4376-25G TCB4376-5G
1-(2-Bromoethoxy)-4-fluorobenzene ≥98.0%
1-(2-Bromoethoxy)-4-fluorobenzene

CAS Number: 332-48-9
MDL Number: MFCD00044739
Molecular Formula: C8H8BrFO
Molecular Weight: 219.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Specific Gravity (20/20): 1.51


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(2-Bromoethoxy)-4-fluorobenzene, 2-(4-Fluorophenoxy)ethyl Bromide, 2-Bromoethyl 4-Fluorophenyl Ether

Formula: C₈H₈BrFO
Boiling Pt: 66 °C
Density: 1.51 (20 °C)
MDL Number: MFCD00044739
CAS Number: 332-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=783a03c56575f9ea9600c0d2adc4ed71723bb6f8&vid=783a03c56575f9ea9600c0d2adc4ed71723bb6f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR