tert-Butyl-3-(isopropyl)carbazate ≥98.0%

Supplier: TCI America

Synonyms: 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-Boc-2-isopropylhydrazine, tert-Butyl 2-isopropylhydrazinecarboxylate

B4375-1G B4375-5G
TCB4375-1GEA 89.27 CAD
TCB4375-1G TCB4375-5G
tert-Butyl-3-(isopropyl)carbazate ≥98.0%
tert-Butyl-3-(isopropyl)carbazate

CAS Number: 16689-35-3
MDL Number: MFCD14635748
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 80
Melting point (°C): 54


Other Product Names / Synonyms: 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 1-Boc-2-isopropylhydrazine

Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Boiling Pt: 80 °C
Melting Pt: 54 °C
MDL Number: MFCD14635748
CAS Number: 16689-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b41d8a3c1a8354c85c58a96c9639b26b71e8a479&vid=b41d8a3c1a8354c85c58a96c9639b26b71e8a479&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR