5-Bromo-2-fluoro-m-xylene ≥97.0% (by GC)
Supplier: TCI America
B4370-1G
B4370-5G
TCB4370-1GEA
46.29
CAD
TCB4370-1G
TCB4370-5G
5-Bromo-2-fluoro-m-xylene ≥97.0% (by GC)
5-Bromo-2-fluoro-m-xylene
CAS Number: 99725-44-7
MDL Number: MFCD01320701
Molecular Formula: C8H8BrF
Molecular Weight: 203.05
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Specific Gravity (20/20): 1.45
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-2-fluoro-1,3-dimethylbenzene
Formula:
C₈H₈BrF Boiling Pt: 89 °C Density: 1.45 (20 °C) |
MDL Number:
MFCD01320701 CAS Number: 99725-44-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...