N-(tert-Butoxycarbonyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Boc-Ida-OH, Boc-DL-Ida-OH, N-Boc-iminodiacetic Acid
B4368-25G
B4368-5G
TCB4368-25GEA
179.96
CAD
TCB4368-25G
TCB4368-5G
N-(tert-Butoxycarbonyl)iminodiacetic acid ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)iminodiacetic acid
CAS Number: 56074-20-5
MDL Number: MFCD00800351
Molecular Formula: C9H15NO6
Molecular Weight: 233.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 126
Storage Temperature: 0-10°C
Other Product Names / Synonyms: N-Boc-iminodiacetic Acid
Formula:
C₉H₁₅NO₆ MW: 233.22 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD00800351 CAS Number: 56074-20-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...