N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-3-fluoro-L-phenylalanine, Boc-Phe(3-F)-OH

B4367-1G B4367-5G
TCB4367-1GEA 72.68 CAD
TCB4367-1G TCB4367-5G
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine

CAS Number: 114873-01-7
MDL Number: MFCD00672522
Molecular Formula: C14H18FNO4
Molecular Weight: 283.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 82
Specific rotation [a]20/D: 7 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-3-fluoro-L-phenylalanine, Boc-Phe(3-F)-OH

Formula: C₁₄H₁₈FNO₄
Melting Pt: 82 °C
MDL Number: MFCD00672522
CAS Number: 114873-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af8deba0c093e44075c57c0a276fa009605d4492&vid=af8deba0c093e44075c57c0a276fa009605d4492&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR