1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%

Supplier: TCI America

Synonyms: 1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

B4364-1G B4364-5G
TCB4364-1GEA 109.02 CAD
TCB4364-1G TCB4364-5G
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester

CAS Number: 761446-45-1
MDL Number: MFCD03789252
Molecular Formula: C16H21BN2O2
Molecular Weight: 284.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 89


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1-Benzylpyrazole-4-boronic Acid Pinacol Ester, 2-(1-Benzyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Formula: C₁₆H₂₁BN₂O₂
MW: 284.16 g/mol
Melting Pt: 89 °C
MDL Number: MFCD03789252
CAS Number: 761446-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=694c5262c33517c05b5dbb286cd6ea39bc98eebe&vid=694c5262c33517c05b5dbb286cd6ea39bc98eebe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR