Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) ≥98.0%

Supplier: TCI America

Synonyms: Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)

B4360-1G B4360-5G
TCB4360-1GEA 319 CAD
TCB4360-1G TCB4360-5G
Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) ≥98.0%
Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II)

CAS Number: 38951-97-2
MDL Number: MFCD10567145
Molecular Formula: C32H28NiO4S4
Molecular Weight: 663.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials), Near Infrared Absorption Materials
Other Product Names / Synonyms: Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)

Formula: C₃₂H₂₈NiO₄S₄
MW: 663.51 g/mol
MDL Number: MFCD10567145
CAS Number: 38951-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e7d80fffbcec188fca056bcba3c014ac812ca8a&vid=3e7d80fffbcec188fca056bcba3c014ac812ca8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR